Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-SMAD2 (Ser245) Polyclonal Antibody
Phospho-SMAD2 (Ser245) Polyclonal Antibody
Supplier: Thermo Scientific PA564515
Description
Phospho-SMAD2 (Ser245) Polyclonal Antibody for Western Blot

Specifications
Phospho-SMAD2 (Ser245) | |
Polyclonal | |
Unconjugated | |
SMAD2 | |
7120426M23Rik; EGK_09664; hMAD-2; hSMAD2; JV18; JV18-1; MAD; MAD homolog 2; Mad protein homolog; Madh2; Madr2; Mad-related protein 2; MGC22139; MGC34440; mMad2; mother against DPP homolog 2; mothers against decapentaplegic homolog 2; Mothers against decapentaplegic-like protein 2; mothers against DPP homolog 2; pSMAD2; Sma- and Mad-related protein 2; SMAD; SMAD 2; SMAD family member 2; SMAD, mothers against DPP homolog 2; Smad2; Smad-2; Smad2Deltaexon3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
17126, 29357, 4087 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O70436, Q15796, Q62432 | |
SMAD2 | |
A synthesized peptide derived from human Smad2 (Phospho-Ser245). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction