Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-Smad2 (Ser465 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE287225UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2872-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2872-25UL Supplier MilliporeSigma Supplier No. ABE287225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-Smad2 (Ser465, Ser467) is a highly specific rabbit polyclonal antibody, that targets Mothers against decapentaplegic homolog 2 and has been tested in Western Blotting, Immunofluorescence and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Smad2 (Ser465
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SMAD2,MADH2,MADR2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 4 amino acids surrounding pSerine from the C-terminus region of human Smad2.
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects SMAD2 phosphorylated on Serine 465 and Serine 467.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.