Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Smad3 (Ser425) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6682 Supplier Bioss Supplier No. BS5616R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMAD3 belongs to the SMAD family of proteins. They are similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.

Specifications

Antigen Phospho-Smad3 (Ser425)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Smad3
Gene Accession No. P84022
Gene Alias AU022421; BOS_10264; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; Madh3; madh3a; madh3-A; MGC60396; mMad3; Mothers against decapentaplegic homolog 3; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; SMAD 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; Smad3a; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3
Gene Symbols Smad3
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human Smad3 around the phosphorylation site of Ser425 [SV (p-S)].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4088
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.