Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-SMO (Ser611, Ser615) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117232
Description
Positive Samples: Jurkat Immunogen sequence: SADVS.
Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.
Specifications
| Phospho-SMO (Ser611, Ser615) | |
| Polyclonal | |
| Unconjugated | |
| SMO | |
| bent body; bnb; E130215L21Rik; frizzled family member 11; FZD11; Gx; HGNC:11119; Protein Gx; Protein SMOH; seven transmembrane helix receptor; SMO; SMOH; smoothened; Smoothened homolog; smoothened homolog (Drosophila); smoothened, frizzled class receptor; smoothened, frizzled family receptor; smoothened, seven transmembrane spanning receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6608 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.08 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99835 | |
| SMO | |
| A synthetic phosphorylated peptide around S611 & S615 of human SMO (NP_0056221). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction