Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPRY4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102834 Supplier Invitrogen™ Supplier No. PA5102834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total prouty 4.

SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRY4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SPRY4
Gene Accession No. Q9C004, Q9WTP2
Gene Alias A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4
Gene Symbols SPRY4
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPRY4(Accession Q9C004), corresponding to amino acid residues L193-V243.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24066, 291610, 81848
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.