Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SRC3 (Ser857) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564836
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564836 Supplier Invitrogen™ Supplier No. PA564836
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-SRC3 (Ser857) Polyclonal Antibody detects endogenous levels of SRC3 only when phosphorylated at Ser857.

AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SRC3 (Ser857)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NCOA3
Gene Accession No. O09000, Q9Y6Q9
Gene Alias 2010305B15Rik; Actr; AIB1; AIB-1; amplified in breast cancer 1 protein; amplified in breast cancer-1 protein homolog; AW321064; BHLHE42; CAGH16; CBP-interacting protein; class E basic helix-loop-helix protein 42; CTG26; KAT13B; MGC141848; NCOA3; NCoA-3; Nuclear receptor coactivator 3; OTTHUMP00000031716; p/Cip; pCip; Rac3; RAC-3; Receptor-associated coactivator 3; RP5-1049G16.3; Src3; SRC-3; steroid receptor coactivator protein 3; Thyroid hormone receptor activator molecule 1; TNRC14; TNRC16; Tram1; Tram-1
Gene Symbols NCOA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NCoA3 (Phospho-Ser857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17979, 8202
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.