Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-SREBP2 (Ser455) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106042 Supplier Invitrogen™ Supplier No. PA5106042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of SREBP2 only when phosphorylated at Ser455.

This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-SREBP2 (Ser455)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SREBF2
Gene Accession No. Q12772, Q3T1I5, Q3U1N2
Gene Alias AI608257; bHLHd2; Class D basic helix-loop-helix protein 2; EMBL:AAI01903.1}; lop13; nuc; Processed sterol regulatory element-binding protein 2; srebf2; srebf2 {ECO:0000312; Srebf2_retired; SREBP2; SREBP-2; SREBP2gc; sterol regulatory element binding factor 2; sterol regulatory element binding protein 2; sterol regulatory element binding transcription factor 2; sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; zgc:158371
Gene Symbols SREBF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SREBF2(Accession Q12772), corresponding to amino acid residues around phosphorylated Ser455.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20788, 300095, 6721
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.