Learn More
Invitrogen™ Phospho-STAT1 (Ser727) Monoclonal Antibody (PSM1)

Description
This antibody recognizes transcriptional factor STAT1 (91 kDa) activated by phosphorylation at Ser727 (intracellular antigen).
Specifications
Specifications
| Antigen | Phospho-STAT1 (Ser727) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PSM1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | STAT1 |
| Gene Accession No. | P42224 |
| Gene Alias | 2010005J02Rik; AA408197; CANDF7; DD6G4-4; DKFZp686B04100; IMD31A; IMD31B; IMD31C; ISGF-3; LOW QUALITY PROTEIN: signal transducer and activator of transcription 1-alpha/beta; OTTHUMP00000165046; OTTHUMP00000205845; signal transducer and activator of transcription 1; signal transducer and activator of transcription 1, 91kD; signal transducer and activator of transcription 1, 91kDa; signal transducer and activator of transcription 1-alpha/beta; signal transducer and activator of transcription-1; STAT; STAT 1; STAT1; STAT-1; stat1 alpha; STAT91; transcription factor ISGF-3; transcription factor ISGF-3 components p91/p84 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.