Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT1 (Y701) Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC2894N100UG
This item is not returnable.
View return policy
Description
STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
STAT1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91 | |
Phosphopeptide containing the human STAT1 Y701 site, Accession # NP009330 | |
100 μg | |
Primary | |
Detects human STAT1 when phosphorylated at Y701 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
1086B | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP009330 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6772 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction