Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT3 (S727) Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4934R100UG
This item is not returnable.
View return policy
Description
STAT3 Monoclonal specifically detects STAT3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| STAT3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6774 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 788335 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Phosphopeptide containing the human STAT3 S727 site | |
| 100 μg | |
| Primary | |
| Detects human STAT3 when phosphorylated at S727. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction