Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT3 (Ser727) Recombinant Rabbit Monoclonal Antibody (SY24-09)

Phospho-STAT3 (Ser727) Recombinant Rabbit Monoclonal Antibody (SY24-09)
Supplier: Thermo Scientific MA532089
Description
Phospho-STAT3 (Ser727) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
Phospho-STAT3 (Ser727) | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P40763, P42227, P52631 | |
STAT3 | |
Synthetic phospho-peptide corresponding to residues surrounding Ser727 of Human STAT3 aa 701-750. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
SY24-09 | |
Unconjugated | |
STAT3 | |
1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 | |
Rabbit | |
Protein A | |
RUO | |
20848, 25125, 6774 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction