Learn More
Phospho-STAT5A/B (Ser726, Ser731) Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | Phospho-STAT5A/B (Ser726, Ser731) |
| Applications | Dot Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 |
| Gene | STAT5B |
| Gene Accession No. | P42229, P51692 |
| Gene Alias | AA959963; Mammary gland factor; mammary gland factor STAT5A; mammary gland factor STAT5B; Mgf; Mpf; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5A; Signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; Stat5a; STAT5B; transcription factor STAT5B |
| Gene Symbols | STAT5A, STAT5B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.