Learn More
Invitrogen™ Phospho-STAT5A/B (Tyr694) Polyclonal Antibody

Description
Positive test controls include: K562. Immunogen sequence: DGYVK.
Specifications
Specifications
| Antigen | Phospho-STAT5A/B (Tyr694) |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.45 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | STAT5A |
| Gene Accession No. | P42229, P42230, P42232, P51692, P52632, Q62771 |
| Gene Alias | AA959963; Mammary gland factor; mammary gland factor STAT5A; mammary gland factor STAT5B; mammary gland factor; transcription factor; MGF; Mpf; signal transducer and activator of transcription 5; Signal transducer and activator of transcription 5a; Signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; Stat5a; Stat5b; transcription factor STAT5B |
| Gene Symbols | STAT5A, STAT5B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.