Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STAT6 (Y641) Rabbit anti-Human, Polyclonal, R&D Systems™
Rabbit Polyclonal Antibody
Supplier: R&D Systems AF3717SP
This item is not returnable.
View return policy
Description
Phospho-STAT6 (Y641) Polyclonal antibody AF3717SP specifically detects Phospho-STAT6 (Y641) in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Simple Western, Western BlottingSpecifications
STAT6 (p Tyr641) | |
Polyclonal | |
Unconjugated | |
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
STAT6 | |
Phosphopeptide containing human STAT6 Y641 site. | |
25 μg | |
Primary | |
Detects human STAT6 when phosphorylated at Y641. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening. |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells | |
D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
Rabbit | |
Affinity Purified | |
RUO | |
6778 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction