Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-STING (Ser365) Polyclonal Antibody
Supplier: Thermo Scientific PA5117271
Description

Specifications
Phospho-STING (Ser365) | |
Polyclonal | |
Unconjugated | |
TMEM173 | |
2610307O08Rik; endoplasmic reticulum IFN stimulator; endoplasmic reticulum interferon stimulator; ERIS; hMITA; hSTING; hypothetical LOC533661; mediator of IRF3 activation; Mita; mitochondrial mediator of IRF3 activation; mitochondrial transmembrane protein 173; MMITA; Mpys; mSTING; NET23; N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner; poSTING; RGD1562552; rSTING; SAVI; Stimulator of interferon genes protein; stimulator of interferon response cGAMP interactor 1; STING; STING1; tm173; Tmem173; Transmembrane protein 173 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
340061, 72512 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TBT3, Q86WV6 | |
TMEM173 | |
A synthetic phosphorylated peptide around S365 of mouse STING (NP_0825371). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction