Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-STK38/STK38L (Thr444, Thr442) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599570 Supplier Invitrogen™ Supplier No. PA599570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of NDR1/2 only when phosphorylated at Thr444/442.

This gene encodes a member of the AGC serine/threonine kinase family of proteins. The kinase activity of this protein is regulated by autophosphorylation and phosphorylation by other upstream kinases. This protein has been shown to function in the cell cycle and apoptosis. This protein has also been found to regulate the protein stability and transcriptional activity of the MYC oncogene. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-STK38/STK38L (Thr444, Thr442)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Stk38
Gene Accession No. Q15208, Q7TSE6, Q91VJ4, Q9Y2H1
Gene Alias 4930473A22Rik; 5830476G13Rik; 9530097A09Rik; AA617404; B230328I19; KIAA0965; NDR; Ndr Ser/Thr kinase-like protein; NDR1; NDR1 protein kinase; NDR2; NDR2 protein kinase; Ndr54; nuclear Dbf2-related 2; nuclear Dbf2-related kinase 1; Nuclear Dbf2-related kinase 2; RGD1564793; serine/threonine kinase 38; serine/threonine kinase 38 like; serine/threonine kinase 38-like; serine/threonine-protein kinase 38; Serine/threonine-protein kinase 38-like; stk38; Stk38l
Gene Symbols Stk38, STK38L
Host Species Rabbit
Immunogen A synthesized peptide derived from human STK38(Accession Q15208), corresponding to amino acid residues around phosphorylated Thr444.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106504, 11329, 23012, 232533, 361813, 691337
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.