Learn More
Invitrogen™ Phospho-STMN2 (Ser73) Polyclonal Antibody

Description
Antibody detects endogenous levels of STMN2 only when phosphorylated at Ser73.
Specifications
Specifications
| Antigen | Phospho-STMN2 (Ser73) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Stmn2 |
| Gene Accession No. | P21818, P55821, Q93045 |
| Gene Alias | AI159727; hypothetical protein LOC534991; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10 |
| Gene Symbols | Stmn2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.