Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-Syk (Tyr323), Rabbit, Alexa Fluor 488, Polyclonal,

Catalog No. ABF1207YMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 488
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABF1207YMI 100 μL Alexa Fluor 488
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF1207YMI Supplier MilliporeSigma™ Supplier No. ABF1207AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

60S ribosomal protein L5 (UniProt P46777; also known as RPL5) is encoded by the RPL5 (also known as DBA6, MSTP030) gene (Gene ID 6125) in human. Ribosomal proteins (RPs) are normally involved in the assembly of the translational machinery-ribosomes essential for protein production. RPL5, RPL11 and the 5S rRNA comprise the 5S ribonucleoprotein particle (RNP), an essential subcomplex of the large ribosomal subunit. RPL5 binds the 5S rRNA, the 5S rRNA/RPL5 complex then localizes to the nucleolus, where it binds RPL11 and is integrated into the ribosome. In response to ribosomal stress (RS) or nucleolar stress, the L5/L11/5S pre-ribosomal complex is redirected to inhibit MDM2 E3 ligase activity, thereby promoting p53 stabilization and p53-dependent cell cycle arrest. RPL5 and RPL11 can also bind TAp73 N-terminal TA domain in an RS-independent manner and thereby prevent MDM2 association with TAp73 target gene promoters, resulting in enhanced TAp73 transcriptional activity and TAp73-dependent apoptosis induction and cell cycle arrest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Syk (Tyr323)
Applications Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Formulation Purified rabbit polyclonal antibody Alexa Fluor™ 488 conjugate in PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene SYK
Gene Accession No. P43405
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence surrounding phosphorylated Tyr323 of human Syk.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003168
Test Specificity Target phosphotyrosine residue coresponds to pTyr323 of human Syk spliced isoform 1 and pTyr300 of human Syk spliced isoform 2. Sequence surrounding the target tyrosine is not well conserved in mouse and rat Syk.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.