Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Syk (Tyr525, Tyr526) Polyclonal Antibody
Phospho-Syk (Tyr525, Tyr526) Polyclonal Antibody
Supplier: Thermo Scientific PA5110162
Description
Phospho-Syk (Tyr525, Tyr526) Polyclonal Antibody for Western Blot, IP

Specifications
Phospho-Syk (Tyr525, Tyr526) | |
Polyclonal | |
Unconjugated | |
Syk | |
DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6850 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P43405 | |
Syk | |
A synthetic phosphorylated peptide around Y525 & Y526 of human Syk (NP_003168.2). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction