Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Syntaxin 1 (Ser14) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105014 Supplier Invitrogen™ Supplier No. PA5105014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Syntaxin 1A only when phosphorylated at Serine 14.

Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Syntaxin 1 (Ser14)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Stx1a
Gene Accession No. O35526, P32851, Q16623
Gene Alias HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2
Gene Symbols Stx1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human STX1A(Accession Q16623), corresponding to amino acid residues around phosphorylated Ser14.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116470, 20907, 6804
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.