Learn More
Invitrogen™ Phospho-TACC3 (Tyr560) Polyclonal Antibody

Description
Antibody detects endogenous levels of TACC3 only when phosphorylated at Tyr560.
Specifications
Specifications
| Antigen | Phospho-TACC3 (Tyr560) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TACC3 |
| Gene Accession No. | Q9JJ11, Q9Y6A5 |
| Gene Alias | Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 |
| Gene Symbols | TACC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.