Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TAK1 (Ser192) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986562
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6562 Supplier Bioss Supplier No. BS5435R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.

Specifications

Antigen Phospho-TAK1 (Ser192)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAP3K7
Gene Accession No. O43318
Gene Alias B430101B05; BOS_9732; C87327; hypothetical protein LOC553788; I79_000697; M3K7; Map3k7; map3k7.L; MAPKKK; MEKK7; mitogen activated protein kinase kinase kinase 7; Mitogen-activated protein kinase kinase kinase 7; mitogen-activated protein kinase kinase kinase 7 L homeolog; RP1-154G14.1; si:ch211-225h9.1; TAK1; TAK1z; TGF1a; TGF-beta activated kinase 1; TGF-beta-activated kinase 1; TGF-beta-activated kinase TAK1; transforming growth factor beta-activated kinase 1; Transforming growth factor-beta-activated kinase 1; XELAEV_18027270mg; xTAK1; zgc:110612
Gene Symbols MAP3K7
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human TAK1 around the phosphorylation site of Ser192.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6885
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.