Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TAK1 (Ser439) Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA540269

Catalog No. PA540269


Only null left
Add to Cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

TAK1 (also known as MAP3K7), a part of the serine/threonine kinase family, is a versatile protein with many signaling functions. Originally identified as a TGF- beta-activated kinase, TAK1 interacts with TGF- beta-receptor and TRAF6 to modulate TGF- beta activation of JNK and p38. It has additional roles in activation of p38 and JNK through Wnt, BMP, and activin signaling pathways as well in response to thyroid hormone, osmotic stress, endothelin, and ephrine. Through activation of von Hippel-Lindau tumor suppressor expression, TAK1 represses PDGF-B, integrin beta1 and integrin beta5, promoting proper wound healing. TAK1 also plays an essential role in IKK activation in numerous signaling pathways including IL-1, IL-6, IL-18, TNF, CD40, TLR, and RIG-I. Activation of the TAK1-IKK pathway requires TAB2/3 and ubiquitination. In this process, TAB2/3 binds to the C-terminal region of TAK1 and becomes polyubiquitinated, TAK1 is autophosphorylated at Thr178, Thr184, Thr187 and Ser192, and finally TAK1 phosphorylates IKK- beta. Additionally, TAK1 represses human telomerase reverse transcriptase suggesting a role in regulation of cell lifespan.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-TAK1 (Ser439)
Polyclonal
Unconjugated
MAP3K7
B430101B05; C87327; hypothetical protein LOC553788; I79_000697; M3K7; map3k7; map3k7.L; MAPKKK; MEKK7; mitogen activated protein kinase kinase kinase 7; mitogen-activated protein kinase kinase kinase 7; mitogen-activated protein kinase kinase kinase 7 L homeolog; RP1-154G14.1; si:ch211-225h9.1; Tak1; TAK1z; TGF1a; TGF-beta activated kinase 1; TGF-beta-activated kinase 1; TGF-beta-activated kinase TAK1; transforming growth factor beta-activated kinase 1; transforming growth factor-beta-activated kinase 1; XELAEV_18027270mg; xTAK1; zgc:110612
Rabbit
Antigen affinity chromatography
RUO
26409, 313121, 6885
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O43318, P0C8E4, Q62073
MAP3K7
Synthesized peptide derived from human Tak1 around the phosphorylation site of S439.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.