Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-Talin (Ser425) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143776
Description
The antibody detects a 240 kDa* protein corresponding to the molecular mass of Talin on SDS-PAGE immunoblots of human A431, rat PC12, and rabbit fibroblast cells treated with Calyculin A, but is not observed after lambda phosphatase treatment.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Specifications
Phospho-Talin (Ser425) | |
Polyclonal | |
Unconjugated | |
TLN2 | |
5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; KIAA1027; mKIAA0320; RGD1565416; talin 1; talin 2; talin-1; Talin-2; TLN; Tln1; Tln2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21894, 313494, 315776, 70549, 7094, 83660 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
P26039, Q71LX4, Q9Y490, Q9Y4G6 | |
TLN1, TLN2 | |
Phospho-Talin (Ser-425) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-425 in human Talin 1. This sequence is conserved in rat, mouse, and chicken Talin 1, as well as in Talin 2 (Ser-428). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction