Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-Tau (Ser235), Mouse anti-Human, Mouse, Clone: RN235, Millipore Sigma™

Catalog No. MABN2275100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2275100 100 μg
MABN227525 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2275100 Supplier MilliporeSigma Supplier No. MABN2275100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Tau (Ser235), clone RN235, Cat. No. MABN2275, is a highly specific mouse monoclonal antibody that targets Microtubule-associated protein tau and has been tested for use in Dot Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.

Microtubule-associated protein tau (UniProt: P10636; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (also known as MAPTL, MTBT1, TAU) gene (Gene ID: 4137) in human. Tau, a highly asymmetric and heat-stable protein, is expressed mainly in the brain, where it regulates the stability and orientation of microtubules in neurons, astrocytes, and oligodendrocytes. Tau protein is reported to be the predominant component of the paired helical filaments (PHFs) and neurofibrillary tangles (NFTs) that are characteristic of pathological brain lesions in Alzheimer s disease. The dephosphorylation of Tau protein promotes rapid and extensive microtubule polymerization, while the phosphorylation of Tau decreases its ability to promote microtubule assembly. Tau is a substrate for a multitude of protein kinases, including CaM kinase II, casein kinase II, protein kinase A (PKA), MAP kinase (ERK2), Cdk5/ p35, and glycogen synthase kinase-3 (GSK-3) and is phosphorylated at many different sites in Alzheimer s disease brain. It is believed that the phosphorylation of specific sites, rather than the overall extent of phosphorylation, is important in modulating the ability of Tau to bind microtubules and promote microtubule assembly. This antibody (clone RN235) specifically reacts with tau phosphorylated at serine 235, and detects intraneuronal NFTs (iNFTs), and extraneuronal NFTs (eNFTs) in the brain tissue. Although theoretically it can also react with phospho Thr231, but EC50 is very high compared to reactivity with phospho Ser235 (407 nM vs 0.61 nM). (Ref.: Brici, D., et al. (2018). J. Alzheimer s Dis. 61(3); 899-905).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Tau (Ser235)
Applications Dot Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RN235
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:500 dilution from a representative lot detected phospho-tau (Ser235) in human Alzheimer's diseas (AD) brain tissue sections.Dot Blot analysis: A representative lot detected phosphorylated recombinant human tau (pSer235), but not the non-phosphorylated recombinant human tau (Tau). (Courtesy of Rebecca Nisbet, Ph.D., University of Queensland, Brisbane, Austraila).Immunofluorescence Analysis: A representative lot detected phospho-tau (Ser235) in K3 and rTg4510 tau transgenic mouse brains (Courtesy of Rebecca Nisbet, Ph.D., University of Queensland, Brisbane, Austraila).Immunohistochemistry (Paraffin) Analysis: A representative lot detected phospho-Tau (Ser235) in human Alzheimer s disease (AD) brain tissue sections. (Courtesy of Rebecca Nisbet, Ph.D., University of Queensland, Brisbane, Austraila).ELISA Analysis: A representative lot full-length human recombinant phospho-tau (Ser235) or GSK-3 beta phosphorylated tau (pTau), but not the non-phosphorylated tau (Courtesy of Rebecca Nisbet, Ph.D., University of Queensland, Brisbane, Austraila).Western Blotting Analysis: A representative lot detected phospho-tau (pSer235) K3 tau transgenic mouse brain homogenates, but not in homogenates from tau knock-out (KO) (Courtesy of Rebecca Nisbet, Ph.D., University of Queensland, Brisbane, Austraila).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Microtubule-associated protein tau;Neurofibrillary tangle protein;Paired helical filament-tau;PHF-tau
Gene Symbols MAPT;MAPTL;MTBT1;TAU
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids surrounding phospho serine 235 of human tau-C.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001116538
Test Specificity Clone RN235 is a mouse monoclonal antibody that detects microtubule-associated protein tau (MAPT) phosphorylated on serine 235 of isoform Tau-C. It targets an epitope with in 12 amino acids surround pSer235. This sequence is also conserved in isoform Tau-F.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.