Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Tau (Thr217) Polyclonal Antibody, HRP, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2843RHRP
This item is not returnable.
View return policy
Description
Phospho-Tau (Thr217) Polyclonal antibody specifically detects Phospho-Tau (Thr217) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| Phospho-Tau (Thr217) | |
| Polyclonal | |
| HRP | |
| MAPT | |
| AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
| KLH conjugated Synthesised phosphopeptide derived from human Tau around the phosphorylation site of Thr217. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| P10636 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4137 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction