Learn More
Phospho-TDP43 (Ser409, Ser410) Mouse anti-Human, Mouse, Rat, Clone: 1A2C1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Phospho-TDP43 (Ser409, Ser410) |
---|---|
Clone | 1A2C1 |
Concentration | 1.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-109-2820
![]() ![]() |
Proteintech Group Inc
663181IG100UL |
100 μL |
Each of 1 for $518.60
|
|
|||||
Description
66318-1-Ig is a phospho-dependent monoclonal antibody specifically recognizing phospho-TDP43 (409/410). It does not react with native TDP-43.
Various biochemical, physiological and behavioural processes display circadian rhythms controlled by an internal biological clock. The central "gears" driving this clock appear to be composed of an autoregulatory transcription/posttranslation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.Specifications
Phospho-TDP43 (Ser409, Ser410) | |
1.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ALS10, phospho(409/410)-TDP43, TAR DNA binding protein, TAR DNA binding protein 43, TARDBP, TDP 43, TDP43 | |
Tardbp | |
IgG1 | |
Protein G | |
Antibody |
1A2C1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q13148, Q921F2 | |
230908, 23435, 298648 | |
Peptide | |
Primary | |
-20°C | |
TARDBP |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.