Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TDP43 (Ser409, Ser410) Recombinant Rabbit Monoclonal Antibody (6M10), Proteintech
SDP

Catalog No. 502333977 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3977 Supplier Proteintech Group Inc Supplier No. 800071RR100UL
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Phospho-TDP43 (Ser409, Ser410) Recombinant Monoclonal antibody specifically detects Phospho-TDP43 (Ser409, Ser410) in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot

Specifications

Antigen Phospho-TDP43 (Ser409, Ser410)
Applications Western Blot
Classification Recombinant Monoclonal
Clone 6M10
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q13148
Gene Alias 1190002A23Rik; ALS10; C85084; TARDBP; TDP-43; TDP43
Host Species Rabbit
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23435
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.