Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TEK (Ser1119) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105922 Supplier Invitrogen™ Supplier No. PA5105922
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TIE2 only when phosphorylated at Ser1119.

The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TEK (Ser1119)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TEK
Gene Accession No. Q02763, Q02858
Gene Alias AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1
Gene Symbols TEK
Host Species Rabbit
Immunogen A synthesized peptide derived from human TEK(Accession Q02763), corresponding to amino acid residues around phosphorylated Ser1119.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21687, 7010
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.