Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho TFEB (Ser142) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABE1971I25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1971-I-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1971-I-25 Supplier MilliporeSigma Supplier No. ABE1971I25UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho TFEB (Ser142) is a highly specific rabbit polyclonal antibody that targets Transcription factor EB phosphorylated on Serine 142 and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho TFEB (Ser142)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols TFEB,BHLHE35
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids surrounding phosphoserine 142 from the N-terminal half of human Transcription factor EB, isoform 1.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001161299
Test Specificity This rabbit polyclonal antibody specifically detects Transcription factor EB phosphorylated on Serine 142 in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.