Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TIE1 (Tyr1117) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105921
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105921 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105921 Supplier Invitrogen™ Supplier No. PA5105921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TIE1 only when phosphorylated at Tyr1117.

Probable protein tyrosine-kinase transmembrane receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TIE1 (Tyr1117)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TIE1
Gene Accession No. P35590, Q06806
Gene Alias D430008P04Rik; JTK14; TIE; Tie1; Tie-1; tyrosine kinase receptor 1; tyrosine kinase with immunoglobulin and epidermal growth factor homology domains 1; tyrosine kinase with immunoglobulin like and EGF like domains 1; tyrosine kinase with immunoglobulin-like and EGF-like domains 1; tyrosine-protein kinase receptor Tie-1
Gene Symbols TIE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TIE1(Accession P35590), corresponding to amino acid residues around phosphorylated Tyr1117.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21846, 7075, 89806
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.