Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-TK1 (Ser13) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538340
Description
Thymidine kinase (TK) belongs to a group of enzymes such as dihydrofolate reductase, thymidylate synthase, and DNA polymerase that are involved in DNA synthesis and precursor production. High levels of these enzymes are present in proliferating cells, and low levels are found in resting cells, due to multiple regulatory mechanisms that ensure exclusive expression of these enzymes in replicating cells. TK is responsible for catalyzing the phosphorylation of thymidine, which functions as a part of the pyrimidine salvage pathway involved in DNA synthesis. The activities of enzymes such as TK are essential for the activation of several chemotherapeutically important nucleoside analogues that are administered as prodrugs.
Specifications
Phospho-TK1 (Ser13) | |
Polyclonal | |
Unconjugated | |
TK1 | |
EC 2.7.1.21, KITH, TK-1, TK1, Thymidine kinase, cytosolic | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21877, 7083 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P04183, P04184 | |
TK1 | |
A synthetic phosphopeptide derived from human TK around the phosphorylation site of Ser13 (P-G-SP-P-S) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction