Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TLK1 (Ser743) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537340 Supplier Invitrogen™ Supplier No. PA537340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 86 kDa. Purity is >95% by SDS-PAGE.

TLK1 or tousled-like kinase 1 is a nuclear serine/threonine kinase that is potentially involved in the regulation of chromatin assembly. TLK1 is first described in Arabidopsis. TLK1 is a novel target of the DNA damage checkpoint that is rapidly inactivated upon exposure to ionizing radiation (IR) and the inactivation is directly mediated by the S-phase DNA damage checkpoint. The suppression of TLK1 activity after double-strand breaks in the DNA, replication blockade, or low doses of ultraviolet irradiation requires ATM, NBS and CHK1. The Tousled-like kinases, first described in Arabadopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TLK1 (Ser743)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TLK1
Gene Accession No. Q8C0V0, Q9UKI8
Gene Alias 4930545J15Rik; E(Sp1) homolog; Enhancer of split groucho-like protein 1; ESG; ESG1; GRG1; KIAA0137; PKU beta; PKU-beta; serine threonine protein kinase; serine/threonine-protein kinase tousled-like 1; SNAK; SNARE protein kinase SNAK; TLE1; TLK1; TLK-1; TLK1B; tousled like kinase 1; tousled-like kinase 1; transducin like enhancer of split 1; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer protein 1
Gene Symbols TLK1
Host Species Rabbit
Immunogen Synthetic phosphopeptide derived from human TLK1 around the phosphorylation site of Serine 743.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228012, 311118, 9874
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.