Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TNK1 (Tyr277) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104671 Supplier Invitrogen™ Supplier No. PA5104671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TNK1 only when phosphorylated at Tyr277.

TNK1 tyrosine kinase, non-receptor 1 belongs to the tyrosine protein kinase family and as such is an important regulator of intracellular signal transduction pathways mediating cellular proliferation, survival, and development. TNK1 is highly expressed in fetal tissues and at lower levels in a few adult tissues. TNK1 may function in signaling pathways utilized broadly during fetal development, and more selectively in adult tissues. TNK1 plays a negative regulatory role in the Ras-Raf1-MAPK pathway, and knockout mice have been shown to develop spontaneous tumors, suggesting a role as a tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TNK1 (Tyr277)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNK1
Gene Accession No. Q13470, Q99ML2
Gene Alias CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1
Gene Symbols TNK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNK1(Accession Q13470), corresponding to amino acid residues around phosphorylated Tyr277.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303247, 83813, 8711
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.