Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TOPBP1 (Ser1138) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114666 Supplier Invitrogen™ Supplier No. PA5114666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TOPBP1 only when phosphorylated at Ser1138.

This gene encodes a binding protein which interacts with the C-terminal region of topoisomerase II beta. This interaction suggests a supportive role for this protein in the catalytic reactions of topoisomerase II beta through transient breakages of DNA strands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TOPBP1 (Ser1138)
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TOPBP1
Gene Accession No. Q6ZQF0, Q92547
Gene Alias 1110031N14Rik; 2810429C13Rik; AI256758; D430026L04Rik; DNA topoisomerase 2-binding protein 1; DNA topoisomerase II-beta-binding protein 1; DNA topoisomerase II-binding protein 1; KIAA0259; mKIAA0259; RGD1562949; TOP2BP1; TOPBP 1; Topbp1; topoisomerase (DNA) II beta binding protein; topoisomerase (DNA) II binding protein 1
Gene Symbols TOPBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TOPBP1 around the phosphorylation site of Ser1138.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11073, 235559, 315969
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.