Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TPH2 (Ser19) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537803 Supplier Invitrogen™ Supplier No. PA537803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is HepG2 cells.

Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TPH2 (Ser19)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Tph2
Gene Accession No. Q8CGU9, Q8CGV2, Q8IWU9
Gene Alias ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Gene Symbols Tph2
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 19(G-F-S(p)-L-D) derived from Human TPH2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121278, 216343, 317675
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.