Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TPH2 (Ser19) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984067
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4067 Supplier Bioss Supplier No. BS17166R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.

Specifications

Antigen Phospho-TPH2 (Ser19)
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tph2
Gene Accession No. Q8IWU9
Gene Alias ADHD7; AU043594; LOC100344397; MGC119994; Neuronal tryptophan hydroxylase; Ntph; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Gene Symbols Tph2
Host Species Rabbit
Immunogen KLH conjugated synthesised phosphopeptide derived from human TPH2 around the phosphorylation site of Ser19.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121278
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.