Learn More
Invitrogen™ Phospho-TRAF2 (Thr22) Polyclonal Antibody

Description
Antibody detects endogenous levels of TRAF2 only when phosphorylated at Thr22.
Specifications
Specifications
| Antigen | Phospho-TRAF2 (Thr22) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TRAF2 |
| Gene Accession No. | P39429, Q12933 |
| Gene Alias | AI325259; E3 ubiquitin-protein ligase TRAF2; MGC:45012; OTTHUMP00000064745; RING-type E3 ubiquitin transferase TRAF2; TNF receptor associated factor 2; Tnf receptor-associated factor 2; TRAF2; TRAP; TRAP3; tumor necrosis factor type 2 receptor associated protein 3; tumor necrosis factor type 2 receptor-associated protein 3 |
| Gene Symbols | TRAF2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.