Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TRIM32 (Ser339) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105349 Supplier Invitrogen™ Supplier No. PA5105349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TRIM32 only when phosphorylated at Ser339.

TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TRIM32 (Ser339)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Trim32
Gene Accession No. Q13049, Q8CH72
Gene Alias 1810045E12Rik; 3f3; 72 kDa Tat-interacting protein; BBS11; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; HT2A; LGMD2H; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; TATIP; Trim32; tripartite motif containing 32; tripartite motif protein 32; tripartite motif-containing 32; tripartite motif-containing protein 32; Zfp117; zinc finger protein 117; zinc finger protein HT2A; zinc-finger protein HT2A
Gene Symbols Trim32
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRIM32(Accession Q13049), corresponding to amino acid residues around phosphorylated Ser339.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22954, 69807
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.