Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TrkA/TrkB (Tyr674, Tyr675, Tyr706, Tyr707) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121285 Supplier Invitrogen™ Supplier No. PA5121285
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa.

This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TrkA/TrkB (Tyr674, Tyr675, Tyr706, Tyr707)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NTRK1
Gene Accession No. P04629, Q16620
Gene Alias BDNF/NT-3 growth factors receptor; BDNF-tropomyosine receptor kinase B; C80751; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; high affinity nerve growth factor receptor; MTC; neural receptor protein-tyrosine kinase (trkB); neurotrophic receptor tyrosine kinase 1; neurotrophic receptor tyrosine kinase 2; Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine receptor kinase type 2; NTRK1; NTRK2; Oncogene TRK; p140-TrkA; RATTRKB1; Slow nerve growth factor receptor; Tkr; Tkrb; trk; TRK1; TRK1-transforming tyrosine kinase protein; Trka; trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TRKB; Trk-B; TrkB tyrosine kinase; TRKB1; Tropomyosin-related kinase A; tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B
Gene Symbols NTRK1, Ntrk2
Host Species Rabbit
Immunogen synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4914, 4915
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.