Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-TrkA (Tyr791) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988573
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8573 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8573 Supplier Bioss Supplier No. BS8456R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TrkA (tropomyosin receptor kinase A) encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. Mutations in the TRKA gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.

Specifications

Antigen Phospho-TrkA (Tyr791)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ntrk1
Gene Accession No. P04629
Gene Alias C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; High affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; NTRK1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; TRKA; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; Tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A
Gene Symbols Ntrk1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human TrkA around the phosphorylation site of Tyr791.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4914
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.