Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TrkC (Tyr705) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599562 Supplier Invitrogen™ Supplier No. PA599562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TrkC only when phosphorylated at Tyr705.

NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TrkC (Tyr705)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ntrk3
Gene Accession No. Q03351, Q16288, Q6VNS1
Gene Alias AW125844; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp145(trkC); GP145-TrkC; neural receptor protein-tyrosine kinase (trkC); neurotrophic receptor tyrosine kinase 3; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; neurotrophin-3; NT-3 growth factor receptor; NTRK3; Ntrk3_tv3; TrkC; trk-C; trkC tyrosine kinase; tyrosine kinase receptor; tyrosine kinase receptor C
Gene Symbols Ntrk3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NTRK3(Accession Q16288), corresponding to amino acid residues around phosphorylated Tyr705.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18213, 29613, 4916
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.