Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tropomyosin-1 alpha (Ser283)Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501992241
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2241 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2241 Supplier Rockland Immunochemicals Supplier No. 600401J52
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Tropomyosin-1 alpha pS283 is directed against the phosphorylated form of humand S283 residue. A BLAST analysis was used to suggest cross-reactivity with mouse and human based on 100% sequence homology. Cross-reactivity with Tropomyosin-1 alpha pS283 from other sources has not been determined.

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.

Specifications

Antigen Phospho-Tropomyosin-1 alpha (Ser283)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and no preservative; pH 7.2
Gene TPM1
Gene Accession No. P09493
Gene Alias AA986836; AI854628; alpha tropomyosin; alpha-fTM; Alpha-tm; Alpha-tropomyosin; alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; BOS_10769; C15orf13; cardiomyopathy, hypertrophic 3; CMD1Y; CMH3; epididymis secretory protein Li 265; HEL-S-265; hepatoma alpha tropomyosin; HTM-alpha; LOC100125989; LVNC9; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; Tm1; TM2; Tm3; Tma2; Tmpa; TMSA; TPM1; Tpm-1; tpm1 protein; TPM1-1; TPM1-alpha; TPMA; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1; tropomyosin 1 (alpha); tropomyosin 1 alpha chain; tropomyosin 1, alpha; tropomyosin 1-1; tropomyosin 3 alpha; Tropomyosin alpha-1 chain; Tropomyosin-1; zgc:171719; zgc:55951; zgc:77009
Gene Symbols TPM1
Host Species Rabbit
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.