Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Troponin I (Ser23, Ser24) Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS3458R

Description
Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. The TnI-fast and TnI-slow genes are expressed in fast-twitch and slow-twitch skeletal muscle fibers, respectively, while the TnI-cardiac gene is expressed exclusively in cardiac muscle tissue. This gene encodes the Troponin-I-skeletal-slow-twitch protein. This gene is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The encoded protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.Specifications
Phospho-Troponin I (Ser23, Ser24) | |
Polyclonal | |
Unconjugated | |
TNNI3 | |
2700018B22Rik; AI747285; BOS_15991; BOS_18114; Cardiac troponin I; cardiac troponin I isoform; cardiomyopathy, dilated 2A (autosomal recessive); CMD1FF; CMD2A; CMH7; Ctni; DKFZp451O223; LOC511094; RCM1; slow skeletal troponin I isoform; SSTNI; Tn1; Tni; TNN1; TNNC1; Tnni1; TNNI3; Tpn1; troponin 1, type 3; troponin I type 1 (skeletal, slow); troponin I type 3 (cardiac); troponin I, cardiac 3; Troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I, skeletal, slow 1; Troponin I, slow isoform; troponin I, slow skeletal; Troponin I, slow skeletal muscle; Troponin I, slow-twitch isoform; troponin I1, slow skeletal type; troponin I3, cardiac type | |
Rabbit | |
Protein A | |
RUO | |
7135, 7137 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P19237, P19429 | |
TNNI1, TNNI3 | |
KLH conjugated synthetic phosphopeptide derived from human Troponin I around the phosphorylation site of Ser23/24. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction