Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TRPM7 (Ser1493) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143778
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143778 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143778 Supplier Invitrogen™ Supplier No. PA5143778
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was cross-adsorbed to unphosphorylated TRPM7 (Ser-1493) then affinity purified using phospho-TRPM7 (Ser-1493) peptide. This antibody detects a 220 kDa* protein on SDS-PAGE immunoblots of human MDA-MB-231 cells stimulated with calyculin A, and detects a 70 kDa autophosphorylated recombinant kinase domain of human TRPM7. These reactivities are not observed after lambda phosphatase treatment to dephosphorylate TRPM7.

TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TRPM7 (Ser1493)
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Trpm7
Gene Accession No. Q96QT4
Gene Alias 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4}
Gene Symbols Trpm7
Host Species Rabbit
Immunogen Phospho-TRPM7 (Ser-1493) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-1493 in human TRPM7. This site is conserved in rat and mouse TRPM7, but the homology surrounding the site is low. This site has no homology to other TRPM family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54822
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.