Learn More
Invitrogen™ Phospho-TPH1 (Ser58) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA14644
Catalog No. PA14644
Only null left
Add to Cart
Description
Description
This antibody is predicted to react with bovine, chicken, human, mouse, non-human primate, Xenopus, rat and zebrafish based on 100% sequence homology. This antibody is specific for the ~55 kDa Tryptophan Hydroxylase protein phosphorylated at Ser58 in Western blots of human dorsal Raphe nucleus.
Tryptophan hydroxylase (TPH) catalyzes the 5-hydroxylation of tryptophan, which is the first step in the biosynthesis of indoleamines (serotonin and melatonin) (Martinez et al., 2001). In mammals, serotonin biosynthesis occurs predominantly in neurons which originate in the Raphe nuclei of the brain, and melatonin synthesis takes place within the pineal gland. Although TPH catalyzes the same reaction within the Raphe nuclei and the pineal gland, TPH activity is rate-limiting for serotonin but not melatonin biosynthesis. Serotonin functions mainly as a neurotransmitter, whereas melatonin is the principal hormone secreted by the pineal gland. The activity of TPH is enhanced by phosphorylation by cAMP-dependent protein kinase (PKA) and Ca2+/calmodulin kinase II (CAM K II) (Jiang et al., 2000; Johansen et al., 1996). Both PKA and CAM K II phosphorylate Ser58 which lies within the regulatory domain of TPH (Kuhn et al., 1997).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-TPH1 (Ser58)
Polyclonal
Unconjugated
TPH1
indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; MGC119994; TPH; Tph1; TPH2; TPRH; TRPH; tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase; tryptophan hydroxylase (tryptophan 5-monooxygenase); tryptophan hydroxylase 1; tryptophan hydroxylase 1 (tryptophan 5-monooxygenase)
Rabbit
Affinity Chromatography
RUO
100009100, 100101558, 21990, 24848
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.3 mg/mL
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P09810, P17290, P17532
TPH1
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser58 of rat TPH1 conjugated to KLH.
100 μL
Primary
Mouse, Rat, Rabbit
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.