Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-TUBA1C (Ser165) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143783 Supplier Invitrogen™ Supplier No. PA5143783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody was cross-adsorbed to unphosphorylated alpha6-Tubulin (a.a. 160-169) peptide before affinity purification using alpha6-Tubulin (Ser-165) peptide (without carrier). The antibody detects a 50 kDa* protein corresponding to the molecular mass of alpha-Tubulin on SDS-PAGE immunoblots of recombinant a1-tubulin phosphorylated with PKCalpha. This reactivity is not observed after lambda phosphatase treatment.

TUBA1C (Tubulin Alpha 1c) is a Protein Coding gene. Diseases associated with TUBA1C include Distal Hereditary Motor Neuropathy, Type Ii. Among its related pathways are Development Slit-Robo signaling and Gap junction. Gene Ontology (GO) annotations related to this gene include GTP binding and structural molecule activity. An important paralog of this gene is TUBA1B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TUBA1C (Ser165)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene TUBA1C
Gene Accession No. P68373, Q6AYZ1, Q9BQE3
Gene Alias alpha-tubulin 6; Alpha-tubulin isotype M-alpha-6; bcm948; Detyrosinated tubulin alpha-1C chain; M[a]6; Tuba1c; TUBA6; tubulin alpha 1c; tubulin alpha-1C chain; Tubulin alpha-6 chain; tubulin, alpha 1C; tubulin, alpha 6
Gene Symbols TUBA1C
Host Species Rabbit
Immunogen Phospho-alpha6-Tubulin (Ser-165) synthetic peptide (coupled to KLH) corresponds to amino acid residues around serine 165 of human alpha6-Tubulin. This sequence is well conserved in most alpha-tubulin isoforms from most eukaryotic species, but is not conserved in beta-Tubulin.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22146, 300218, 84790
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.