Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-TYK2 (Tyr1054) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537762
Description
A suggested positive control for Western blot is HT29 cells.
Tyrosine Kinase 2 (TYK2) a member of the JAK kinase family, plays both structural and catalytic roles in type I interferon (IFN) signaling. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.
Specifications
Phospho-TYK2 (Tyr1054) | |
Polyclonal | |
Unconjugated | |
TYK2 | |
4833430L21Rik; AW540162; BRE1; BRE1A; C79397; hBRE1; IMD35; JTK1; mKIAA4116; Non-receptor tyrosine-protein kinase TYK2; Tyk2; tyrosine kinase 2; tyrosine kinase TYK2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100361294, 54721, 7297 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P29597, Q9R117 | |
TYK2 | |
Peptide sequence around phosphorylation site of tyrosine 1054 (H-E-Y(p)-Y-R) derived from Human TYK2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction