Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Tyrosine Agarose-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems AGM1676
This item is not returnable.
View return policy
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Immunoprecipitation.Specifications
Phospho-Tyrosine | |
Monoclonal | |
Agarose | |
Phosphate-buffered saline (PBS) with preservative. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects endogenous proteins containing phosphorylated Tyrosine residues. ELISA and 2D Western blot analyses using ligand- and pervanadate-treated cell lysates indicate that clone 179003 binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. Does not cross-react with proteins or peptides containing phosphorylated serine or threonine residues. | |
Multi-species | |
Purified |
Immunoprecipitation | |
179003 | |
Immunoprecipitation 10 uL/500 ug cell lysate | |
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine | |
KLH-coupled phospho-tyrosine synthetic peptide. | |
250 μL | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction