Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Tyrosine Alkaline Phosphatase-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. APM1676
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APM1676 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. APM1676 Supplier R&D Systems Supplier No. APM1676
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Phospho-Tyrosine
Applications Western Blot, ELISA
Classification Monoclonal
Clone 179003
Conjugate Alkaline Phosphatase (AP)
Dilution Western Blot 1:1000 dilution, ELISA
Formulation Supplied as a 0.2 μm filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives. with No Preservative
Gene Alias 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine
Host Species Mouse
Immunogen KLH-coupled Phospho-Tyrosine synthetic peptide.
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects proteins containing phosphorylated Tyrosine residues. ELISA and 2D Western blot analyses using pervanadate-treated cell lysates indicate that it binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. This antibody does not cross-react with proteins or peptides containing phosphorylated serine or threonine residues.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Multi-species
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.